Phosphate, Low Range
Supplier: LaMotte
Certificates
About this item
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
- Range: 0–2 ppm
- Material for 50 Tests
- Compact, Waterproof Carrying Case Contains Kit
- Instructions Included
- Pre-calibrated Test Tubes
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.