Phacops rana milleri (Devonian)
Catalog #: 470237-644
Supplier: Avantor
Specifications
- Description:Phacops rana milleri (Devonian) Fossil Reproduction
- Cat. no.:470237-644
Specifications
About this item
Dramatically preserved assemblage of 24 complete trilobites showing exceptional detail on a 13×15" matrix. Ohio.
Customers Who Bought This Also Bought