Peridinium Slide
Catalog #: 470181-020
Supplier: Avantor
Specifications
- Description:Peridinium
- Cat. no.:470181-020
- Stain:Fast Green and Mallory's
- Supplier No.:WARD470181-020
- Orientation:Whole mount
Specifications
About this item
Plated Cell Walls, Two Flagella
- Identifying features clearly distinguishable
- Rigorous quality control standards
Freshwater. High concentrations cause odor issues in drinking water.
Customers Who Bought This Also Bought