Ward's® Peranema
Catalog #: 470177-302
Supplier: Ward's Science
Specifications
- Description:Peranema (Small)
- No. of Students:30
- Cat. no.:470177-302
- Packaging:Jar
- Media:Hay culture medium
- Supplier No.:WARD470177-302
Specifications
About this item
Extremely small euglenoid. Usually doesn’t rotate when swimming. Scavenger. Hay medium.
Characterized by flagella; lacks plastids.
Customers Who Bought This Also Bought