Peat Moss
Catalog #: 470006-560
Supplier: INDUSTRIAL SUPPORT INC.
Small Business Enterprise
Specifications
- Description:Peat Moss
- Cat. no.:470006-560
- Supplier No.:6563400
Specifications
About this item
Increase acidity and promote moisture retention with this soil mixer, which can help you start seeds, rootings, transplants and cuttings. 1.35kg.
Customers Who Bought This Also Bought