Peak Loupe
Catalog #: 470021-810
Supplier: Selsi
Specifications
- Magnification:15×
- Description:Peak Loupe
- Cat. no.:470021-810
- Supplier No.:1962
Specifications
About this item
Large Lenses for Wide, Distortion-Free Viewing Area
The clear plastic base of this convenient loupe can be placed over a live specimen to contain it for closer examination. Size: 1 1/4"L x 1 3/8" widest diameter. 15X magnification.
Customers Who Bought This Also Bought