Paneth Cells Slide
Catalog #: 470182-868
Supplier: Avantor
Specifications
- Description:Paneth Cells
- Cat. no.:470182-868
- Stain:Phloxine/tartrazine
- Supplier No.:WARD470182-868
- Orientation:Section
Specifications
About this item
Cells Found Throughout the Small Intestine
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Stained to show Paneth cells in red.
Customers Who Bought This Also Bought