Osmosis Apparatus Set
Catalog #: 470005-200
Supplier: Avantor
Specifications
- Description:Osmosis Apparatus Set
- Cat. no.:470005-200
- Supplier No.:WARD470005-200
Specifications
About this item
Insert a sugar solution into the 16" clear tube, lower the membrane end into the jar of water, and watch the water level rise.
Customers Who Bought This Also Bought