Ward's® Live Oscillatoria Culture
Supplier: Ward's Science
Certificates
About this item
Oscillatoria is a genus of filamentous green algae that moves by sliding filaments back and forth against each other.
- Filamentous, Freshwater green algae
- Unbranched trichome, highly sheathed
- Genus gets its name from the oscillating motion it exhibits
Genus: Oscillatoria Species: not specified
Related Information
Algae_and_Cyanobacteria.pdf Working_with_Algae_and_Cyanobacteria.pdf
Product Literature
Customers Who Bought This Also Bought