Specifications
- Description:Wards® Chemistry Teacher Demonstration: Oscillating Yellow and Blue Solution
- Cat. no.:470010-824
- Includes:Instructions
- Supplier No.:WARD470010-824
- No. of Uses:5
Specifications
About this item
Teach Basic Chemical Concepts in a Fun, Visual Way.
- Can be performed in just 10–30 minutes
- Requires only basic labware that you provide
For this demonstration you simply place a flask containing a yellow solution on a magnetic mixer and watch the solution turn from yellow to blue and back to yellow. The reaction will continue to oscillate for 10–15 minutes. Give your students a visual representation of a redox reaction and teach them about the chemical reaction concepts while enjoying the color changes.