Grafco® Opthalmoscope
Catalog #: 470019-304
Supplier: Graham-Field
Specifications
- Description:Opthalmoscope Set
- Cat. no.:470019-304
- Supplier No.:1227-2
Specifications
About this item
Examine the interior of the eye.
- The set features a dioptic range of –20 to +20
The pinhole opening emits light based on the diopter scale, and the chrome-plate, set-screw lock handle with rheostat houses two C batteries.
Ordering information: C batteries has to be ordered separately.
Delivery: Supplied with replacement bulb and case.
Customers Who Bought This Also Bought