Order Entry
ContactUsLinkComponent
Oleic Acid
Oleic Acid
Catalog #: 470301-906
Supplier:  Ward's Science
CAS Number:  
OLEIC ACID LG 500ML
Oleic Acid
Catalog #: 470301-906
Supplier:  Ward's Science
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Oleic Acid
  • Size:
    500 mL
  • Chemical/Physical Specifications:
    Solution
  • Cat. no.:
    470301-906
  • Grade:
    Laboratory
  • Supplier No.:
    OX0010-500ML
  • Disposal Method:
    This disposal method is published for your convenience. You MUST have checked with your federal, state, and local regulations before using this method and these methods are only applicable for small laboratory sized quantities.

    Use a lab apron, chemical resistant gloves and goggles before following the below method to dispose of hazardous waste.

     1. To a large beaker, add a large volume of water in a fume hood.
     2. Add the organic acid to the beaker.
     3. Neutralize the mixture by adding dilute sodium hydroxide or sodium carbonate until the pH is about 7. Stir.
     4. Pour the final solution down the drain with at least a 20-fold excess of water.

Specifications

About this item

CAS Number: 112-80-1
Formula Weight: 282.47
Formula: CH3(CH2)7CH:CH(CH2)7COOH
Density (g/mL): 0.895
Boiling Point (°C): 286
Freezing Point (°C): 11-14
Solubility: Organic Solvents, Not Water
Synonyms: Oleinic Acid, Red Oil, cis-9-Octadecanoic Acid
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Customers Who Bought This Also Bought