Octahedrite
Catalog #: 470026-534
Supplier: ADAM MOORE MO
Specifications
- Description:Octahedrite
- Cat. no.:470026-534
- Supplier No.:495720
Specifications
About this item
Polished, etched iron meteorite slice showing Widmanstatten structure, 1–1 1/2".
Customers Who Bought This Also Bought