Nutrient Agar Deeps, Tubes
Catalog #: 470180-660
Supplier: Avantor
CAS Number:
Specifications
- Description:Prepared Nutrient Agar Deeps, Tubes
- Cat. no.:470180-660
- Supplier No.:WARD470180-660
Specifications
About this item
Contains 12 nutrient agar deeps. Used to check for organism motility. 17 mL per tube.
Customers Who Bought This Also Bought