Nitrate-Nitrogen
Supplier: LaMotte
Certificates
About this item
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
- Range: 0 to 15 ppm
- Material for 50 tests
- Compact, waterproof carrying case contains kit
- Instructions included
- Pre-calibrated test tubes
- Aligned with GLOBE protocols
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.