Navicula
Catalog #: 470179-846
Supplier: Ward's Science
Specifications
- Description:Navicula pelliculosa (Small)
- No. of Students:30
- Cat. no.:470179-846
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470179-846
Specifications
About this item
Freshwater. Unicellular; pennate, with bilateral symmetry. Basic culture solution.
Characterized by shells (tests) made of silica, in two parts.
Customers Who Bought This Also Bought