Ward's® Mouse Maze
Catalog #: 470019-324
Supplier: Dynalon
Specifications
- Description:Mouse Maze
- Cat. no.:470019-324
- Dimensions:61×61×9 cm
- Supplier No.:8551-2663
Specifications
About this item
Removable walls let you change the configuration of the maze.
- Perfect for studying the behavior of rodents and other small animals
This maze with removable walls allows students to set up any pattern they wish. The clear acrylic cover allows students to observe the animals progress without any danger of escape. Made of polypropylene, this durable maze is ideal for use with all age groups and is perfect for science fair students for years to come. The maze may also be put into a dishwasher or autoclave to remove trace smells and pheromones left behind.
Customers Who Bought This Also Bought