TeacherGeek Motor Adaptor Pin
Catalog #: 470132-442
Supplier: TEACHERGEEK INC
Specifications
- Description:Motor Adaptor Pin, Pkg. of 90
- Cat. no.:470132-442
- Supplier No.:27619
Specifications
About this item
TeacherGeek Motor Adaptor Pins slide onto the motor shaft and fit securely in the splined hole of a small gear or pulley.
TeacherGeek Motor Adaptor Pins slide onto the motor shaft and fit securely in the splined hole of a small gear or pulley. This allows you to easily motorize an activity or original design.
This allows you to easily motorize an activity or original design.
Customers Who Bought This Also Bought