Specifications
- Description:Mini-Prep Isolation of Plasmid DNA
- Cat. no.:470121-614
- Supplier No.:202-B
Specifications
About this item
Allow students to practice small-scale rapid isolation of plasmid DNA, which is a routine procedure used for screening and analysis of recombinant DNA in cloning and subcloning experiments. In this experiment, students isolate plasmid DNA in just 60 minutes, without the use of toxic chemicals such as phenol or chloroform.
Material provided can support up to 40 isolations and five DNA agarose gels. Kit includes instructions, Plasmid Extraction LyphoCells, gel loading solution, UltraSpec-Agarose powder, electrophoresis buffer, InstaStain FlashBlue and FlashBlue Plus stain, calibrated pipet, 100 ml graduated cylinder and microtipped transfer pipettes. Time requirements: 60 min isolation of plasmid; 45 min electrophoresis.