Specifications
- Description:MimioPad™ wireless tablet
- Cat. no.:470017-942
- Supplier No.:1747666
Specifications
About this item
Work on the Whiteboard From a Distance
Add the MimioPad™ wireless tablet to your MimioTeach™ system, and you and your students can collaborate on whiteboard lessons from up to 30 feet away from the classroom computer. The MimioPad tablet works with a slim pen stylus powered by a rechargeable Lithium-Ion battery and included MimioStudio software. You can access the most common operations of MimioStudio software from custom-tailored shortcut keys on the tablet. Plus, up to 50 MimioPad tablets can connect to one computer, so students can participate without leaving their seats. Package includes MimioPad tablet, stylus, USB mini-B cable (0.9m/3 ft), USB RFwireless receiver, pen tip extractor tool, two replaceable pen tips, rechargeable Lithium-Ion type battery, optional pen tether, and three CDs containing system installers. A two year warranty on all hardware components can be extended to five years total with product registration online or by mail. Limited lifetime warranty available on software.