Microscopy Skills Slide Set
Catalog #: 470177-054
Supplier: Avantor
Specifications
- No. of Slides:3
- Description:Microscopy Skills Slide Set
- Cat. no.:470177-054
- Supplier No.:WARD470177-054
Specifications
About this item
Learn Focusing Skills and Understand Scale
- Identifying features clearly distinguishable
- Rigorous quality control standards
All slides are prepared as whole mounts.
Ordering information: Set includes Cross colored threads; Letter “e”; Stage micrometer.
Customers Who Bought This Also Bought