Micro, Pure Gum Amber Rubber Tubing
Catalog #: 470150-362
Supplier: Kent
Specifications
- Description:Micro, Pure Gum Amber Rubber Tubing
- Cat. no.:470150-362
- Supplier No.:202-12
Specifications
About this item
Small bore with light wall, surgical grade. ID: ¹/₁₆"; OD: ¹/₈"; wall thickness: ¹/₃₂". 12' (3.6 m) length.
Customers Who Bought This Also Bought