Mica Set
Catalog #: 470024-862
Supplier: Avantor
Specifications
- Description:Mica Set
- Cat. no.:470024-862
- Supplier No.:WARD470024-862
Specifications
About this item
Specially selected thin sections of important rock forming minerals. Set may contain more than one example of the same mineral to depict variations in occurrence and distinct mineral associations.
Slides (12); annite, biotite, clinochlore, fuschite, glauconite, lepidolite, margarite, muscovite, paragonite, phlogopite, sericite, zinnwaldite.
Customers Who Bought This Also Bought