Metric Micrometer, United Scientific Supplies
Catalog #: 470005-644
Supplier: United Scientific Supplies
Specifications
- Measurement range (°Brix):0-25 mm
- Measurement range:0-25 mm
- Resolution:0.01 mm
- Cat. no.:470005-644
- Supplier No.:MCR025
Specifications
About this item
Excellent Quality
- Precise
- Forged steel frame
Ratchet stop ensures equal pressure on parts being measured.
Customers Who Bought This Also Bought