Metal Cube and Slab Set
Catalog #: 470104-974
Supplier: American Scientific
Small Business Enterprise
Specifications
- Description:Metal cube and slab set
- Cat. no.:470104-974
- Supplier No.:CEM-225
Specifications
About this item
A steel cube, an aluminum cube and a rectangular aluminum slab have each been painted to conceal the identity of the metal. The steel and aluminum cubes have equal volume while the aluminum slab has greater volume. One plastic metric ruler is included with this set. For use with IPS textbook. Set of three.
WARNING: CHOKING HAZARD - Small parts. Not for children under 3 years.
Customers Who Bought This Also Bought