Medium Wall Red Rubber Tubing, Machine Made
Supplier: Longwood Elastomers
Certificates
Certificates
About this item
An economical, smooth finish tubing which is stiffer than the handmade type. Machine made by extrusion.
- Specific gravity: 1.23
Customers Who Bought This Also Bought
Nalgene® Laboratory Notebooks, Thermo Scientific
Supplier: Thermo Fisher Scientific
172,57 $ to 531,06 $
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
1 747,88 $
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
541,80 $
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
401,76 $
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
121,35 $ to 993,13 $