Marble Ramp
Catalog #: 470003-992
Supplier: Avantor
Specifications
- Description:Marble Ramp
- Cat. no.:470003-992
- Supplier No.:WARD470003-992
Specifications
About this item
Ideal for students conduct experiments by varying the angle of the marble track as well as the height from which the marble is released.
- Includes instructions
- Grades 7 - 9
- Size: 96.5 x 12.7cm
By gathering data and plotting a graph, students discover that the same result can often be achieved by different combinations of angle and height.
Caution: CHOKING HAZARD - Item contains a marble. Not for children under 3 yrs.
Customers Who Bought This Also Bought