Magni-Grip Magnifier with Forceps
Catalog #: 470021-818
Supplier: CARSON OPTICAL
Specifications
- Magnification:4×
- Description:Magni-Grip Magnifier with Forceps
- Cat. no.:470021-818
- Supplier No.:MG-55
Specifications
About this item
Perform Fine Dissections and Handle Small Specimens Easily
The 4X magnifying lens is attached to fine-tipped stainless steel forceps to view even very small specimens. The forceps act as a base for the magnifier for hands-free observation. It folds for storage. Size: 4 1/4"L x 1 1/2"W, lens size 1 1/8" dia., forceps size 3 1/4"L.
Customers Who Bought This Also Bought