Liquid Cyanoacrylate Glue
Catalog #: 470024-626
Supplier: FORENSICS SOURCE
Specifications
- Description:Glue, 1 oz.
- Cat. no.:470024-626
- Supplier No.:LL-0002
Specifications
About this item
This glue is suitable for use in forensics.
- Produces hard, white visible impressions
- 1 oz.
When heated in a closed chamber, the fumes from this “Super Glue” adhesive react with the residues in latent prints to produce hard, white visible impressions.
Customers Who Bought This Also Bought