Lilium Ovary, Second Four-Nucleate Embryo Sac Slide
Catalog #: 470181-266
Supplier: Avantor
Specifications
- Description:Lilium Ovary, Second Four-Nucleate Embryo Sac
- Cat. no.:470181-266
- Stain:Quadruple stained
- Orientation:Cross section
Specifications
About this item
Angiosperm Reproduction
- Identifying features clearly distinguishable
- Rigorous quality control standards
Lily. In series.
Customers Who Bought This Also Bought