Light Source
Catalog #: 470111-326
Supplier: Norman Lamps
Specifications
- Description:Light Source, Replacement Bulb
- Cat. no.:470111-326
- Supplier No.:7.5S11-13VFR
Specifications
About this item
A handy addition to your optical bench, this light source features a 7.5 watt incandescent lamp plus a cord and plug for connection to 110 VAC power source.
Customers Who Bought This Also Bought