To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- ā¢ State issued document with your organization's Federal Tax ID Number
- ā¢ State issued document with your organization's Resale Tax ID Number
- ā¢ City or County issued Business License
- ā¢ State Department of Health Services License
- ā¢ Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:LaMotte Soil pH TesTabs Kit
- Cat. no.:470021-532
- Supplier No.:5912
- No. of Tests:50
Specifications
About this item
An Ideal Alternative to Liquid pH Indicators
- No hazardous compounds required for testing
- Colorimetric pH determination
- Includes instructions
Quickly and simply measure soil pH with these non-hazardous reagents. The kit includes 50 tests and a laminated color chart for comparisons and pH determinations. Chart values feature range 4.0 to 11.0 pH. Distilled or deionized water recommended. Includes instructions.