Ward's® Lac Operon: Turning On Your Genes Activity Lab Activity
Catalog #: 470004-742
Supplier: Avantor
Specifications
- Description:Lac Operon: Turning On Your Genes Activity Kit
- No. of Lab Groups:15
- Cat. no.:470004-742
- Supplier No.:WARD470004-742
Specifications
About this item
Useful for teaching AP Biology units on Molecular Genetics.
- Illustrate multiple types of gene regulation
- Teacher developed and classroom tested
This manipulative activity can be used to show how genes are "turned off and on" to prevent waste of energy and resources, as well as to demonstrate concepts of proteins, enzymes, and gene expression. Students label and define all parts of the operon and walk through the steps of the operon when it is considered on and off. This can then be tied into lessons on positive and negative feedback loops. Developed by Carol Bullock. Includes materials for 30 students with accompanying DVD and instructions.
Customers Who Bought This Also Bought