Stainless Steel Laboratory Scoops
Supplier: PRECISION MOULDED POLYMERS LIMITED
Certificates
About this item
Excellent stainless steel scoops with wooden handle.
One end has a point for handling small amounts of chemicals and the other end is blunt. The scoop is 16.5 cm long. The 14-cm handle is made of varnished hardwood.
Customers Who Bought This Also Bought