LaMotte® Iron Water Test Kit
Supplier: LaMotte
Certificates
About this item
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
- Range: 0.5 to 10 ppm
- Materials for 50 tests
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.