Iron Meteorite
Catalog #: 470026-532
Supplier: ADAM MOORE MO
Specifications
- Description:Iron Meteorite
- Specimen Dimensions:1 - 1½"
- Supplier No.:495715
Specifications
About this item
Fragments in natural condition, many showing concave forms and fusion crusts. Sizes are approximate.
- Available in two sizes
- Slicked in oil to prevent rusting
- Protected in bubble wrap and a sealed bag
Customers Who Bought This Also Bought