Specifications
- Description:Ionic Interaction Lab Activity
- No. of Lab Groups:6
- Cat. no.:470007-768
- Supplier No.:4-104
Specifications
About this item
This kit provides an introduction to ionic interactions by giving students exposure to a large number of reactions and allowing maximum observation for a limited amount of time and effort.
- Students Write Ionic Formulas and Equations
- Lab Includes Follow-Up Activities
- Test Can Be Repeated 100 Times
- Ideal for High School Chemistry or Physical Science Studies
- Activities Help Students Use Empirical Evidence to Draw Conclusions
It includes precipitate forming, gas forming, and non-ionized product forming reactions. The investigation may be extended to teach students a number of additional basic chemical skills such as writing ionic formulas for the products formed, writing ionic interaction equations, relating placement of elements in the periodic table to their reactivity, and developing solubility principles from empirical evidence.
This kit is ideal for high school chemistry or physical sciences studies.