Lab-Aids® Investigating Human Respiration Kit
Supplier: Lab-Aids
Certificates
About this item
Explore the role Of respiratory system in regulation of blood gases.
- Creating models is one way to understand and communicate scientific information
- Examine human respiration
- Time required: one to two 50 minutes class periods
Using bromothymol blue (BTB) as an indicator, students conduct a qualitative investigation. The amount of carbon dioxide in their exhaled breath is then quantitatively measured using the indicator to perform a simple titration. The combination of these steps demonstrates the function of the lungs in providing surface area for oxygen to enter and carbon dioxide wastes to leave the blood.
Delivery: See datasheet for contents information.
Customers Who Bought This Also Bought