Kemtec® Introduction to Toxicology
Catalog #: 470144-334
Supplier: Kemtec
Specifications
- Description:Introduction to toxicology
- No. of Lab Groups:6
- Cat. no.:470144-334
- Supplier No.:10-530
Specifications
About this item
Learn how to screen for drugs.
- Practice thin layer chromatography
- Learn about polarity and pH
- Time required: 50 minute class period
Acquaint your students with the variety of procedures used in testing unknowns against known standards for the presence of narcotics. Techniques employed include TLC, testing polarity/pH of molecules, urinalysis, and more. The activity comes with enough materials for six setups and a teacher and student manual.
Ordering information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Delivery: See datasheet for contents information.
Customers Who Bought This Also Bought