Specifications
- Description:Introduction to Biochemistry 1
- No. of Lab Groups:6
- Cat. no.:470144-432
- Supplier No.:2-216
Specifications
About this item
Explore basic biochemistry.
- For biology, general science, chemistry or biochemistry courses
- All required chemicals included
- Target grades 8-12
This kit was designed to introduce students to the chemistry of living things and relate biochemistry to everyday life. Includes five basic biochemistry experiments in which students use chemical methods to test for proteins, carbohydrates and lipids, use Pepsin to digest proteins, and Ninhydrin to detect amino acids, test the activity of salivary amylase (both in vitro and in vivo), develop a technique for determining ascorbic acid concentration and use it to evaluate the vitamin C levels in substances, and determine the alkalinity and acidity of foods. A great exploratory or introductory experience is gained by performing the test procedures.
Includes:
Instructor Manual
SDS
Methods of Assessment Manual
pH paper 1-14
Pipettes
Filter Paper
Forceps
Indophenol
Ninhydrin
Pepsin
Starch
Protein
Ascorbic Acid
Glucose
Sudan Solution
Benedict's Solution
Biuret Solution
Acetone Solution
Vegetable Oil
Potassium Solution
Hydrochloric Acid Solution
Teacher's Guide and Student Copymaster