Specifications
- Description:Interferometer
- Cat. no.:470121-282
- Supplier No.:U10350
Specifications
About this item
Complete both Michelson interferometer and Fabry-Perot interferometer experiments with this high-grade equipment set.
Complete both Michelson interferometer and Fabry-Perot interferometer experiments with this high-grade equipment set. With the optional accessory kit, students can also determine the refractive index of glass, refractive index of air, and Twyman-Green qualitative test for optical components. This complete equipment set of optical components is mounted on a heavy, rigid base plate for precise and reproducible measurements. The large optical components permit a generation of clear and well-defined interference patterns in daylight. The reflective observation screen also features an adjustable inclination. Pre-defined component positions allow for quick rearrangement to ensure extremely rapid preparation for the various experiments. The equipment set includes a stable plastic box for storing the mounted and adjusted interferometer as well as the laser base plate.
Beam splitter: Evenness: λ/10 (front side), λ/4 (rear side) Size: 40 mm dia. Surface-coated mirror: Evenness: \<λ/2. Size: 40 x 40 mm2. Eccentric reduction: approx. 1:1000 (individual calibration specified on eccentric base) Size: 24.5 x 33 x 2.5cm; weight: 10 lbs. A He-Ne Laser and the Accessory Kit, both available separately are required to complete some of the experiments.
Interferometer Accessory Kit
This accessory set for the interferometer consists of a vacuum cell for determining the refractive index of air and a glass plate on a rotatable holder for examining the surface quality of optical components or Twyman-Green interferometer. A vacuum hand pump and silicone tube are also required.
- Large Optical Components
- Well Defined Interference Patterns
- Eccentric Reduction: approximately 1:1000
- Stable, Plastic Storage Box
- Rigid Base Plate
This complete equipment set of optical components is mounted on a heavy, rigid base plate for precise and reproducible measurements. The large optical components permit a generation of clear and well-defined interference patterns in daylight. The reflective observation screen also features an adjustable inclination. Pre-defined component positions allow for quick rearrangement to ensure extremely rapid preparation for the various experiments. The equipment set includes a stable plastic box for storing the interferometer and the laser base plate.
The beam splitter has an evenness of λ/10 (front side), λ/4 (rear side) and measures 40 mm dia. Surface-coated mirror has an evenness of λ/2 and is 40 x 40 mm. The eccentric reduction is approx. 1:1000 (individual calibration specified on eccentric base) and is 2.5 x 24.5 x 33 cm. It weighs 10 lb. A vacuum hand pump and silicone tube are required.