Specifications
- Description:Wind to Hydrogen Science Kit
- No. of Lab Groups:1
- Cat. no.:470039-908
- Supplier No.:FCJJ-26
Specifications
About this item
The Wind to Hydrogen Science Education Kit enables students to invent their own clean energy applications using a small electric motor powered by a fuel cell.
Explore how the power generated through the use of wind and water make great alternative energy solutions with this engaging kit. Students will build their own Hydrogen-powered inventions and be able to visibly see the generated wind power in the form of illuminated LED lights and musical sounds. The kit includes a miniature real-working turbine which is designed for students to evaluate the pitch of the profiled blades. Also included is a complete curriculum on wind energy and fuel cells with easy to follow experiment manual, assembly guide, and background history on the technology.
Restriction Comments:
This science kit should only be used by persons 12 years old and up, and only under the supervision of adults who have familiarized themselves with the safety measures described in the science kit. Warning: Choking Hazard: Not suitable for children under 3 yrs. The fuel cell generates gases that are very easily ignited.
- Contains a hydrogen fuel cell and wind turbine
- Electric motor can be used for various applications
- 12 years old and up
Hydrogen is produced through water electrolysis by a reversible PEM fuel cell that is itself powered by a wind turbine with profiled blades based on NASA aeronautics. This hydrogen is then converted to electrical energy by the fuel cell and can be used to power a small fan.