M12 Complete™ Electrophoresis Package
Catalog #: 470103-996
Supplier: Edvotek
Specifications
- Description:M12 Horizontal Dual Electrophoresis Cell
- Cat. no.:470103-996
- Supplier No.:504
Specifications
About this item
Run the full spectrum of horizontal electrophoresis experiments with this versatile package!
- Sleek New Design Speeds Electrophoresis
- Includes Complete Set of New & Improved Electrophoresis Accessories
- Now Includes a DNA DuraGel™ (Cat. #S-43) for Pipetting Practice!
- Large Color Coded Push Tabs for Easy Lid Insertion & Removal
- Pour Spout for Buffer Disposal
- Improved Ventilation Reduces Lid Condensation
- User Replaceable Electrodes
- Chamber Reverse Compatible with Previous Edvotek® Accessories
- US Design Patent No. D749,235
- Made in USA
Our newly reimagined M12 Complete™ supports one or two student groups in two standard length gel trays for experiments that require less separation, or one long gel tray for experiments that require more. Produces excellent results in 15-30 minutes and includes a lifetime warranty.
Delivery: Includes: (2) 7×7 cm Gel Trays, (1) 7×14 cm Gel Tray, (1) Duragel, (2) 6 Tooth Combs, (2) 8/10 Tooth Combs, (4) Rubber End Caps.
Customers Who Bought This Also Bought