Hooded Gas Lighters
Catalog #: 470005-604
Supplier: G C Fuller
Specifications
- Description:Hooded gas lighter
- Cat. no.:470005-604
- Supplier No.:S1501
Specifications
About this item
Produce a cean, economical, and safe flame.
- Design ensures spark ignites combustible vapor only
- Hood trap gas (for positive ignition) and shields the striking file
Customers Who Bought This Also Bought