Specifications
- Description:Historical Geology Reference Set
- Cat. no.:470026-806
- Supplier No.:WARD470026-806
- No. of Parts:45 samples
Specifications
About this item
Teacher’s Master Set for Use with Student GEO-logic® Historical Geology Sets
- Developed by teachers and WARD's Staff Geologists
- Classroom tested for dependabilty
- Designed to save valuble planning and preparation time
Ideal for studying topics such as types of fossils and methods of fossilization, and drawing conclusions about the Earth’s history. This GEO-logic set comes with a teacher’s guide with suggested class activities that include classifying fossils by type of fossilization; plant, animal, or “in between”; symmetry in fossils; identifying fossils by key; morphology — clue to recognition; and fossils as clues to the Earth’s history. The set is housed in a compartmented collection box.
Contents:
45 numbered fossils; magnifierFossils:Algae (3), Protozoa (3), Porifera (sponges) (2), Cnidaria (corals) (3), Bryozoa (sea mats) (3), Brachiopoda (lamp shells) (4), Scaphopoda (1), Pelecypoda (clams, oysters) (6), Gastropoda (snails) (3), Cephalopoda (2), Annelida (worm) (1), Trilobita (4), Echinodermata (4), Graptoloidea (1), Chordata (3), Plants (2)
The GEO-logic system provides a modular approach to teaching and allows you to address broad curriculum needs for the study of minerals and rocks; natural resources and energy; and fossils and earth history. The two types of GEO-logic teaching collections include: A Reference Set, which provides an overview of all topics and comes with a teacher’s guide. Each GEO-logic collection includes: Individually numbered specimens chosen for their quality and character, Activity guides with learning objectives and hands-on activities for your students to perform, and an attractive, durable compartmented box for easy use and storage.