Specifications
- Description:Glow Chemistry Kit II: It’s Blue!
- Cat. no.:470105-492
- Supplier No.:WARD470105-492
Specifications
About this item
Chemistry teacher, Stephen Freedman, uses this demonstation at Halloween.
- Grades 8 to 12
Two solutions, A and B, are created with ammonium bicarbonate, hydrogen peroxide, sodium bicarbonate, sodium carbonate luminol and copper II sulfate. Mix the solutions together and a dramatic blue glow appears, which is very eerie, especially in a darkened room.