Ward's® Live Gloeocapsa Culture
Supplier: Ward's Science
Certificates
About this item
This colonial cyanobacteria is structured as several cells bound together in a gelatinous "mucilaginous sheath".
Commonly found in masses on wet rocks in aquatic or semi-aquatic habitats.
Genus: Gloeocapsa Species: Not specified
Related Information
Algae_and_Cyanobacteria.pdf Working_with_Algae_and_Cyanobacteria.pdf
Product Literature
Customers Who Bought This Also Bought