GLOBE® Soil Color Book
Catalog #: 470005-702
Supplier: Peco Sales
Specifications
- Description:GLOBE Soil Color Book
- Cat. no.:470005-702
- Supplier No.:77369
Specifications
About this item
Created especially to meet the requirements of the GLOBE program.
- Aligned with GLOBE protocols
The color chips in this spiral-bound book are specially formulated using more earth-based pigments to make it easier to match under less than perfect lighting conditions. Over 200 chips and a soil structure key are included in the set. Size: 7 1/4" x 3 1/4".
Customers Who Bought This Also Bought