GLOBE Cloud Chart
Catalog #: 470104-448
Supplier: Peco Sales
Specifications
- Description:GLOBE Cloud Chart
- Cat. no.:470104-448
- Supplier No.:33485
Specifications
About this item
Clouds cover the world and this chart, written in English, French, Spanish, Arabic, Chinese and Russian make that point very well indeed.
- Size: 11×15"
Provides descriptions and visual examples of the ten basic cloud types: cirrus, cirrostratus, cirrocumulus, altostratus, altocumulus, cumulus, nimbostratus, stratus, cumulonimbus, and stratocumulus.
Customers Who Bought This Also Bought