Giant Color Coded Periodic Table
Catalog #: 470016-786
Supplier: BRITTEN BANNER INC.
Specifications
- Width:96”
- Description:Giant Color Coded Periodic Table
- Height:48”
- Supplier No.:4751300
Specifications
About this item
Large enough to be seen from ANY corner of the classroom.
- Also available in 8.5 x 11" student pads and transparency
- Uses IUPAC and older CAS numbering
This oversized periodic table clearly displays all elements, including synthetic and radioactive elements. Color coding makes it easy for your students to distinguish between metals, nonmetals, metalloids and groups. Table uses both the new IUPAC group numbering system (1–18) and the older CAS numbering system (IA, IIA, IIIB, VIB...). Details given for each element include name, symbol, atomic number, atomic weight, and a state of matter at 20°C. Also included is a table of naturally occurring isotopes and a color coded table of common uses.
Customers Who Bought This Also Bought